siRNA Details
| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317915.1 | NP_001304844.1 | 5 | 7264 | ||
| NM_138453.3 | NP_612462.1 | 5 | 7226 | |||
| GenBank mRNA | AB073648.1 | - | 1 | 396 | ||
| AK055409.1 | - | 1 | 427 | |||
| AK094685.1 | - | 1 | 1273 | |||

RAB3C
Genomic Map