siRNA Details
| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317915.1 | NP_001304844.1 | 5 | 6103 | ||
| NM_138453.3 | NP_612462.1 | 5 | 6065 | |||
| GenBank mRNA | AK094685.1 | - | 1 | 112 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB3C| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317915.1 | NP_001304844.1 | 5 | 6103 | ||
| NM_138453.3 | NP_612462.1 | 5 | 6065 | |||
| GenBank mRNA | AK094685.1 | - | 1 | 112 | ||