siRNA Details
| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317915.1 | NP_001304844.1 | 5 | 7467 | ||
| NM_138453.3 | NP_612462.1 | 5 | 7429 | |||
| GenBank mRNA | AB073648.1 | - | 1 | 599 | ||
| AB074188.1 | - | 1 | 424 | |||
| AK055409.1 | - | 1 | 630 | |||
| AK094685.1 | - | 1 | 1476 | |||

RAB3C
Genomic Map