siRNA Details
| Gene Symbol: CNTN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125798.1 | - | 2 | 488 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB061357.1 | - | 2 | 488 | Chr. 3 - 1008141 - 1009787 [+] on Build GRCh38 | |

CNTN6
Genomic Map