siRNA Details
| Gene Symbol: LOC105369705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093502.1 | - | 1 | 1736 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369705| Gene Symbol: LOC105369705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093502.1 | - | 1 | 1736 | ||