siRNA Details
| Gene Symbol: LOC105375092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056771.1 | - | 1 | 1151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105375092| Gene Symbol: LOC105375092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056771.1 | - | 1 | 1151 | ||