siRNA Details
| Gene Symbol: LOC105375092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056771.1 | - | 1 | 164 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA837430.1 | - | 1 | 165 | Chr. 6 - 53065824 - 53066385 [-] on Build GRCh38 | |

LOC105375092
Genomic Map