siRNA Details
Gene Symbol: ZNF366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152625.2 | NP_689838.1 | 5 | 5353 | ||
GenBank mRNA | AK090694.1 | - | 1 | 1563 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152625.2 | NP_689838.1 | 5 | 5353 | ||
GenBank mRNA | AK090694.1 | - | 1 | 1563 |