siRNA Details
| Gene Symbol: ZNF366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152625.2 | NP_689838.1 | 5 | 6095 | ||
| GenBank mRNA | AK090694.1 | - | 1 | 2305 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF366| Gene Symbol: ZNF366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152625.2 | NP_689838.1 | 5 | 6095 | ||
| GenBank mRNA | AK090694.1 | - | 1 | 2305 | ||