siRNA Details
| Gene Symbol: LINC01498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120483.1 | 2 | 270 | |||
| GenBank mRNA | BX648662.1 | - | 6 | 1473 | ||
| DB089475.1 | - | 2 | 270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01498| Gene Symbol: LINC01498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120483.1 | 2 | 270 | |||
| GenBank mRNA | BX648662.1 | - | 6 | 1473 | ||
| DB089475.1 | - | 2 | 270 | |||