siRNA Details
| Gene Symbol: LOC101927027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110204.1 | 5 | 1647 | |||
| GenBank mRNA | CN415424.1 | - | 1 | 546 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK128542.1 | - | 1 | 4149 | Chr. 2 - 178435401 - 178440059 [+] on Build GRCh38 | |

LOC101927027
Genomic Map