siRNA Details
| Gene Symbol: KCTD21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC062331.1 | - | 1 | 933 | 5798762 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD21-AS1| Gene Symbol: KCTD21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC062331.1 | - | 1 | 933 | 5798762 | |