siRNA Details
| Gene Symbol: LOC284112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098506.1 | - | 1 | 2421 | ||
| Gene Symbol: MIR497HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038310.1 | 2 | 427 | |||

LOC284112
Genomic Map