siRNA Details
| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047701.1 | 8 | 2982 | |||
| NR_135580.1 | 8 | 1392 | ||||
| NR_135583.1 | 7 | 2917 | ||||
| XM_005249807.4 | XP_005249864.1 | 7 | 2981 | |||
| XM_006715752.2 | XP_006715815.1 | 8 | 2949 | |||
| XM_017012414.1 | XP_016867903.1 | 7 | 2908 | |||
| XM_017012415.1 | XP_016867904.1 | 8 | 3010 | |||
| XM_017012416.1 | XP_016867905.1 | 7 | 2908 | |||
| XM_017012417.1 | XP_016867906.1 | 8 | 1412 | |||
| XM_017012418.1 | XP_016867907.1 | 7 | 1282 | |||
| XM_017012421.1 | XP_016867910.1 | 8 | 3132 | |||
| XM_017012422.1 | XP_016867911.1 | 8 | 3021 | |||
| XM_017012424.1 | XP_016867913.1 | 7 | 2924 | |||
| XM_017012430.1 | XP_016867919.1 | 8 | 1384 | |||
| GenBank mRNA | AK001665.1 | - | 8 | 2952 | ||
| AK023007.1 | - | 9 | 2491 | |||
| AK023053.1 | - | 8 | 1324 | |||
| AK127933.1 | - | 2 | 1912 | |||

COA1
Genomic Map