siRNA Details
| Gene Symbol: LOC101928524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125127.1 | - | 1 | 1263 | ||
| BX647538.1 | - | 1 | 3101 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928524| Gene Symbol: LOC101928524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125127.1 | - | 1 | 1263 | ||
| BX647538.1 | - | 1 | 3101 | |||