siRNA Details
| Gene Symbol: LOC101928524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055017.1 | - | 1 | 508 | ||
| BX647538.1 | - | 1 | 4167 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA060916.1 | - | 1 | 508 | Chr. 10 - 13642305 - 13642854 [+] on Build GRCh38 | |

LOC101928524
Genomic Map