siRNA Details
| Gene Symbol: LINC01484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK074818.1 | - | 2 | 939 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01484| Gene Symbol: LINC01484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK074818.1 | - | 2 | 939 | ||