siRNA Details
Gene Symbol: LINC01180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110661.1 | 2, 3 | 1685 | |||
GenBank mRNA | BC043289.1 | - | 2, 3 | 1689 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110661.1 | 2, 3 | 1685 | |||
GenBank mRNA | BC043289.1 | - | 2, 3 | 1689 |