siRNA Details
| Gene Symbol: LOC105372795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131189.1 | 1 | 607 | |||
| GenBank mRNA | BC062470.1 | - | 2 | 546 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ719284.1 | - | 2 | 545 | Chr. 21 - 37063038 - 37064040 [-] on Build GRCh38 | |

LOC105372795
Genomic Map