siRNA Details
| Gene Symbol: LOC105372795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131189.1 | 1 | 883 | |||
| GenBank mRNA | BC062470.1 | - | 2 | 822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105372795| Gene Symbol: LOC105372795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131189.1 | 1 | 883 | |||
| GenBank mRNA | BC062470.1 | - | 2 | 822 | ||