siRNA Details
Gene Symbol: LOC105372795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131189.1 | 1 | 336 | |||
GenBank mRNA | BC062470.1 | - | 1 | 311 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105372795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131189.1 | 1 | 336 | |||
GenBank mRNA | BC062470.1 | - | 1 | 311 |