siRNA Details
| Gene Symbol: SNAI3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094837.1 | - | 1 | 73 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA254069.1 | - | 1 | 73 | Chr. 16 - 88673396 - 88673986 [+] on Build GRCh38 | |

SNAI3-AS1
Genomic Map