siRNA Details
| Gene Symbol: OR2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516019.2 | XP_011514321.1 | 3 | 491 | ||
| XM_011516020.2 | XP_011514322.1 | 3 | 266 | |||
| GenBank mRNA | AJ459868.1 | - | 2 | 173 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2F1| Gene Symbol: OR2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516019.2 | XP_011514321.1 | 3 | 491 | ||
| XM_011516020.2 | XP_011514322.1 | 3 | 266 | |||
| GenBank mRNA | AJ459868.1 | - | 2 | 173 | ||