siRNA Details
| Gene Symbol: ADAM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003812.3 | NP_003803.1 | 26 | 5290 | ||
| XM_005246932.3 | XP_005246989.1 | 25 | 5299 | |||
| XM_011512086.2 | XP_011510388.1 | 26 | 5390 | |||
| GenBank mRNA | AF052115.1 | - | 1 | 741 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX114973.1 | - | 1 | 416 | Chr. 2 - 206619780 - 206620533 [+] on Build GRCh38 | |

ADAM23
Genomic Map