siRNA Details
| Gene Symbol: ADAM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003812.3 | NP_003803.1 | 26 | 5739 | ||
| XM_005246932.3 | XP_005246989.1 | 25 | 5748 | |||
| XM_011512086.2 | XP_011510388.1 | 26 | 5839 | |||
| GenBank mRNA | AF052115.1 | - | 2 | 1159 | ||

ADAM23
Genomic Map