siRNA Details
| Gene Symbol: TIAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126861.1 | - | 1 | 642 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124161.1 | - | 1 | 2192 | Chr. 17 - 29076584 - 29078857 [-] on Build GRCh38 | |

TIAF1
Genomic Map