siRNA Details
| Gene Symbol: SSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025253.1 | XP_016880742.1 | 15 | 1730 | ||
| GenBank mRNA | AF086010.1 | - | 1 | 129 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSH2| Gene Symbol: SSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025253.1 | XP_016880742.1 | 15 | 1730 | ||
| GenBank mRNA | AF086010.1 | - | 1 | 129 | ||