siRNA Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006713560.2 | XP_006713623.1 | 16 | 7421 | ||
| XM_011512612.2 | XP_011510914.1 | 14 | 10675 | |||
| XM_011512613.1 | XP_011510915.1 | 14 | 7483 | |||
| XM_011512615.2 | XP_011510917.1 | 13 | 10532 | |||
| XM_011512616.2 | XP_011510918.1 | 13 | 8213 | |||
| XM_017006070.1 | XP_016861559.1 | 18 | 7863 | |||
| XM_017006071.1 | XP_016861560.1 | 19 | 7933 | |||
| XM_017006072.1 | XP_016861561.1 | 20 | 8067 | |||
| XM_017006073.1 | XP_016861562.1 | 18 | 7666 | |||
| XM_017006074.1 | XP_016861563.1 | 20 | 10085 | |||
| XM_017006075.1 | XP_016861564.1 | 17 | 8810 | |||
| XM_017006076.1 | XP_016861565.1 | 17 | 7603 | |||
| XM_017006077.1 | XP_016861566.1 | 19 | 9014 | |||
| XM_017006078.1 | XP_016861567.1 | 17 | 8831 | |||
| XM_017006079.1 | XP_016861568.1 | 15 | 8307 | |||
| XM_017006080.1 | XP_016861569.1 | 15 | 8678 | |||
| XM_017006081.1 | XP_016861570.1 | 13 | 10528 | |||
| XM_017006082.1 | XP_016861571.1 | 15 | 8526 | |||
| XM_017006083.1 | XP_016861572.1 | 14 | 6967 | |||
| GenBank mRNA | AK021754.1 | - | 1 | 280 | ||

NAALADL2
Genomic Map