siRNA Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006713560.2 | XP_006713623.1 | 16 | 8493 | ||
| XM_011512612.2 | XP_011510914.1 | 14 | 11747 | |||
| XM_011512613.1 | XP_011510915.1 | 14 | 8555 | |||
| XM_011512615.2 | XP_011510917.1 | 13 | 11604 | |||
| XM_011512616.2 | XP_011510918.1 | 13 | 9285 | |||
| XM_017006070.1 | XP_016861559.1 | 18 | 8935 | |||
| XM_017006071.1 | XP_016861560.1 | 19 | 9005 | |||
| XM_017006072.1 | XP_016861561.1 | 20 | 9139 | |||
| XM_017006073.1 | XP_016861562.1 | 18 | 8738 | |||
| XM_017006074.1 | XP_016861563.1 | 20 | 11157 | |||
| XM_017006075.1 | XP_016861564.1 | 17 | 9882 | |||
| XM_017006076.1 | XP_016861565.1 | 17 | 8675 | |||
| XM_017006077.1 | XP_016861566.1 | 19 | 10086 | |||
| XM_017006078.1 | XP_016861567.1 | 17 | 9903 | |||
| XM_017006079.1 | XP_016861568.1 | 15 | 9379 | |||
| XM_017006080.1 | XP_016861569.1 | 15 | 9750 | |||
| XM_017006081.1 | XP_016861570.1 | 13 | 11600 | |||
| XM_017006082.1 | XP_016861571.1 | 15 | 9598 | |||
| XM_017006083.1 | XP_016861572.1 | 14 | 8039 | |||
| GenBank mRNA | AK021754.1 | - | 1 | 1352 | ||

NAALADL2
Genomic Map