siRNA Details
| Gene Symbol: LRPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002337.3 | NP_002328.1 | 8 | 2447 | ||
| NR_110005.1 | 8 | 2325 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC006574.1 | - | 2 | 671 | Chr. 4 - 3509185 - 3512409 [-] on Build GRCh38 | |

LRPAP1
Genomic Map