siRNA Details
| Gene Symbol: GPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY358864.1 | - | 6 | 2406 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124793.1 | - | 1 | 1974 | Chr. 1 - 26876133 - 26878169 [-] on Build GRCh38 | |

GPN2
Genomic Map