siRNA Details
| Gene Symbol: KCNK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104210.1 | 3 | 2058 | |||
| XM_011517101.2 | XP_011515403.1 | 3 | 1813 | |||
| XM_011517102.2 | XP_011515404.1 | 3 | 4009 | |||
| XM_011517103.2 | XP_011515405.1 | 3 | 1557 | |||
| XM_017013530.1 | XP_016869019.1 | 3 | 3725 | |||
| XM_017013531.1 | XP_016869020.1 | 3 | 1824 | |||
| GenBank mRNA | AK090707.1 | - | 2 | 1491 | ||
| AK095204.1 | - | 1 | 279 | |||
| BC034734.1 | - | 2 | 904 | |||

KCNK9
Genomic Map