siRNA Details
| Gene Symbol: LINC01108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108096.1 | 3 | 2740 | |||
| NR_108097.1 | 2 | 2504 | ||||
| GenBank mRNA | AK056826.1 | - | 2 | 2195 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU186607.1 | - | 1 | 37 | Chr. 6 - 14280228 - 14281120 [-] on Build GRCh38 | |

LINC01108
Genomic Map