siRNA Details
Gene Symbol: LINC01108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108096.1 | 3 | 2500 | |||
NR_108097.1 | 2 | 2264 | ||||
GenBank mRNA | AK056826.1 | - | 2 | 1955 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108096.1 | 3 | 2500 | |||
NR_108097.1 | 2 | 2264 | ||||
GenBank mRNA | AK056826.1 | - | 2 | 1955 |