siRNA Details
| Gene Symbol: LINC01108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108096.1 | 3 | 2778 | |||
| NR_108097.1 | 2 | 2542 | ||||
| GenBank mRNA | AK056826.1 | - | 2 | 2233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01108| Gene Symbol: LINC01108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108096.1 | 3 | 2778 | |||
| NR_108097.1 | 2 | 2542 | ||||
| GenBank mRNA | AK056826.1 | - | 2 | 2233 | ||