siRNA Details
| Gene Symbol: IQCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095094.1 | - | 2 | 2837 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK096574.1 | - | 1 | 1063 | Chr. 3 - 197955287 - 197958085 [-] on Build GRCh38 | |

IQCG
Genomic Map