siRNA Details
| Gene Symbol: DBN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534447.2 | XP_011532749.1 | 1 | 801 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL117546.1 | - | 1 | 89 | Chr. 5 - 177476631 - 177479656 [-] on Build GRCh38 | |

DBN1
Genomic Map