siRNA Details
| Gene Symbol: IBA57-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103539.1 | 2 | 275 | |||
| NR_103540.1 | 2 | 119 | ||||
| GenBank mRNA | BX114038.1 | - | 2 | 275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IBA57-AS1| Gene Symbol: IBA57-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103539.1 | 2 | 275 | |||
| NR_103540.1 | 2 | 119 | ||||
| GenBank mRNA | BX114038.1 | - | 2 | 275 | ||