siRNA Details
Gene Symbol: IBA57-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103539.1 | 1 | 177 | |||
GenBank mRNA | BX114038.1 | - | 1 | 177 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK057440.1 | - | 1 | 518 | Chr. 1 - 228164125 - 228165014 [-] on Build GRCh38 |
