siRNA Details
| Gene Symbol: IBA57-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103539.1 | 2 | 539 | |||
| NR_103540.1 | 2 | 383 | ||||
| GenBank mRNA | BX114038.1 | - | 2 | 538 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK057440.1 | - | 1 | 1041 | Chr. 1 - 228164125 - 228165014 [-] on Build GRCh38 | |

IBA57-AS1
Genomic Map