siRNA Details
Gene Symbol: ASTN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033973.1 | 10 | 1368 | |||
GenBank mRNA | AK057447.1 | - | 10 | 1368 | ||
AK097822.1 | - | 1 | 91 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB054482.1 | - | 1 | 91 | Chr. 9 - 116552066 - 116568639 [+] on Build GRCh38 |
