siRNA Details
| Gene Symbol: SLC38A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX641065.1 | - | 6 | 2203 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK057666.1 | - | 1 | 974 | Chr. 5 - 55633021 - 55635083 [-] on Build GRCh38 | |

SLC38A9
Genomic Map