siRNA Details
| Gene Symbol: ZNF654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL357201.1 | - | 1 | 341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF654| Gene Symbol: ZNF654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL357201.1 | - | 1 | 341 | ||