siRNA Details
Gene Symbol: FAM95C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047651.1 | 2, 3 | 2391 | |||
GenBank mRNA | AK097951.1 | - | 2, 3 | 2393 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM95C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047651.1 | 2, 3 | 2391 | |||
GenBank mRNA | AK097951.1 | - | 2, 3 | 2393 |