siRNA Details
| Gene Symbol: FAM95C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047651.1 | 3 | 2519 | |||
| GenBank mRNA | AK097951.1 | - | 3 | 2521 | ||
| AL834388.1 | - | 1 | 772 | |||
| Gene Symbol: ANKRD18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093059.1 | - | 8 | 2275 | ||

FAM95C
Genomic Map