siRNA Details
| Gene Symbol: FAM95C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047651.1 | 2 | 2022 | |||
| GenBank mRNA | AK097951.1 | - | 2 | 2024 | ||
| Gene Symbol: ANKRD18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093059.1 | - | 7 | 1065 | ||
| BC131497.1 | - | 7 | 1162 | |||

FAM95C
Genomic Map