siRNA Details
| Gene Symbol: LOC105374809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL832916.1 | - | 1 | 231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374809| Gene Symbol: LOC105374809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL832916.1 | - | 1 | 231 | ||