siRNA Details
| Gene Symbol: TMEM132B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286219.1 | NP_001273148.1 | 5 | 7752 | ||
| NM_052907.3 | NP_443139.2 | 9 | 9005 | |||
| XM_011537852.2 | XP_011536154.1 | 9 | 9062 | |||
| XM_011537854.2 | XP_011536156.1 | 5 | 13735 | |||
| XM_017018767.1 | XP_016874256.1 | 10 | 9171 | |||
| GenBank mRNA | AK094786.1 | - | 1 | 763 | ||

TMEM132B
Genomic Map