siRNA Details
| Gene Symbol: TMEM132B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286219.1 | NP_001273148.1 | 5 | 8583 | ||
| NM_052907.3 | NP_443139.2 | 9 | 9836 | |||
| GenBank mRNA | AK094786.1 | - | 1 | 1594 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM132B| Gene Symbol: TMEM132B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286219.1 | NP_001273148.1 | 5 | 8583 | ||
| NM_052907.3 | NP_443139.2 | 9 | 9836 | |||
| GenBank mRNA | AK094786.1 | - | 1 | 1594 | ||