siRNA Details
| Gene Symbol: LINC01361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110632.1 | 3 | 803 | |||
| NR_110633.1 | 3 | 800 | ||||
| GenBank mRNA | BC037222.1 | - | 3 | 803 | ||
| U80773.1 | - | 3 | 800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01361| Gene Symbol: LINC01361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110632.1 | 3 | 803 | |||
| NR_110633.1 | 3 | 800 | ||||
| GenBank mRNA | BC037222.1 | - | 3 | 803 | ||
| U80773.1 | - | 3 | 800 | |||